MilliporeSigma
  • Plasminogen Improves Mouse IVF by Interactions with Inner Acrosomal Membrane-Bound MMP2 and SAMP14.
Plasminogen Improves Mouse IVF by Interactions with Inner Acrosomal Membrane-Bound MMP2 and SAMP14.
Biology of reproduction (2016-03-05)
Marvin J S Ferrer, Wei Xu, Jagathpala Shetty, John Herr, Richard Oko
ABSTRACT
Spermatozoa must penetrate the outer investments of the oocyte, the cumulus oophorus and the zona pellucida (ZP), in order for fertilization to occur. This may require exposure of enzymes on the sperm's inner acrosomal membrane (IAM), one of which is matrix metalloproteinase (MMP) 2, to factors in oviductal fluid. Plasminogen is present in oviductal fluid and activates MMP2 in somatic tissues. The objectives of this study were: 1) to examine possible interactions between plasminogen and IAM-bound plasminogen activator receptor (SAMP14) and -MMP2, 2) to demonstrate plasminogen's presence in the extracellular environment at the site of fertilization, and 3) to provide evidence that plasminogen plays a role in fertilization. Zymographs of sonicated bull and rat sperm extracts incubated with plasmin and/or plasminogen (plasmin/ogen) showed acceleration of initiation of MMP2 activity in concentrations as low as 1 μg/ml. Immunohistochemical and immunofluorescence analysis of plasmin/ogen revealed its presence in the cytoplasm of mouse ovarian and oviductal oocytes, oviductal epithelium, around the ZP, and amongst the cumulus cells. We modified the standard in vitro fertilization (IVF) approach to more closely mimic natural fertilization by reducing sperm concentration during insemination by ∼100× and also comparing cumulus-intact and denuded oocytes. In mice, addition of plasminogen in IVF medium significantly improved fertilization, while MMP2 antibody significantly inhibited sperm penetration in these conditions. IVF improvement by plasminogen was blocked by SAMP14 antibody. Furthermore, MMP2 antibody inhibition was coincident with a failure by spermatozoa to disperse the cumulus oophorus. We provide evidence that plasminogen on its own and through an MMP2-related mechanism improves the ability of oocytes to be fertilized, and demonstrate its effect in sperm penetration of oocyte investments.
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~10,000 IU