MilliporeSigma
  • Preparation of cyclodextrin-modified monolithic hybrid columns for the fast enantioseparation of hydroxy acids in capillary liquid chromatography.
Preparation of cyclodextrin-modified monolithic hybrid columns for the fast enantioseparation of hydroxy acids in capillary liquid chromatography.
Journal of separation science (2016-03-31)
Kamila Szwed, Junjie Ou, Guang Huang, Hui Lin, Zhongshan Liu, Hongwei Wang, Hanfa Zou
ABSTRACT
Cyclodextrins and their derivatives are one of the most common and successful chiral selectors. However, there have been few publications about the use of cyclodextrin-modified monoliths. In this study, organic hybrid monoliths were prepared by the immobilization of derivatized β-cyclodextrin alone or with l-2-allylglycine hydrochloride to the polyhedral oligomeric silsesquioxane methacryl substituted monolith. The main topic of this study is a combined system with dual chiral selectors (l-2-allylglycine hydrochloride and β-cyclodextrin) as monolithic chiral stationary phase. The effect of l-2-allylglycine hydrochloride concentration on enantioseparation was investigated. The enantioseparation of the four acidic compounds with resolutions up to 2.87 was achieved within 2.5 min on the prepared chiral monolithic column in capillary liquid chromatography. Moreover, the possible mechanism of enantioseparation was discussed.
Sigma-Aldrich
10-Undecenoyl chloride, 97%