MilliporeSigma
  • Design and evaluation of lipoprotein resembling curcumin-encapsulated protein-free nanostructured lipid carrier for brain targeting.
Design and evaluation of lipoprotein resembling curcumin-encapsulated protein-free nanostructured lipid carrier for brain targeting.
International journal of pharmaceutics (2016-04-21)
Fanfei Meng, Sajid Asghar, Yurui Xu, Jianping Wang, Xin Jin, Zhilin Wang, Jing Wang, Qineng Ping, Jianping Zhou, Yanyu Xiao
ABSTRACT
Many nanoparticle matrixes have been demonstrated to be efficient in brain targeting, but there are still certain limitations for them. To overcome the shortcomings of the existing nanoparticulate systems for brain-targeted delivery, a lipoprotein resembling protein-free nanostructured lipid carrier (PS80-NLC) loaded with curcumin was constructed and assessed for in vitro and in vivo performance. Firstly, single factor at a time approach was employed to investigate the effects of various formulation factors. Mean particle sizes of ≤100nm, high entrapment efficiency (EE, about 95%) and drug loading (DL, >3%) were obtained for the optimized formulations. In vitro release studies in the presence of plasma indicated stability of the formulation under physiological condition. Compared with NLC, PS80-NLC showed noticeably higher affinity for bEnd.3 cells (1.56 folds greater than NLC) but with lower uptake in macrophages. The brain coronal sections showed strong and widely distributed fluorescence intensity of PS80-NLC than that of NLC in the cortex. Ex vivo imaging studies further confirmed that PS80-NLC could effectively permeate BBB and preferentially accumulate in the brain (2.38 times greater than NLC). The considerable in vitro and in vivo performance of the safe and biocompatible PS80-NLC makes it a suitable option for further investigations in brain targeted drug delivery.
Product Description
Trilaurin, European Pharmacopoeia (EP) Reference Standard