MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Evidence for OTUD-6B participation in B lymphocytes cell cycle after cytokine stimulation.
Evidence for OTUD-6B participation in B lymphocytes cell cycle after cytokine stimulation.
PloS one (2011-01-27)
Zhongping Xu, Yufang Zheng, Yufei Zhu, Xiangyin Kong, Landian Hu
ABSTRACT
Deubiquitinating enzymes (DUBs) are important regulators of cell proliferation. Here we identified a functional deubiquitinating enzyme, ovarian tumor domain-containing 6B (OTUD-6B). Mutation of the conserved Cys residue abolished its deubiquitinating activity in vitro. Otud-6b expression was induced with cytokine stimulation in both mouse Ba/F3 cells and primary B lymphocytes followed a rapid decrease. This rapid decrease was partially facilitated by tristetraprolin (TTP) destabilization of Otud-6b mRNA through AU-rich motifs. Enforced expression of OTUD-6B in Ba/F3 cells could block cell proliferation by arresting cells in G1 phase. In addition, cyclin D2 level was down-regulated when OTUD-6B WT was overexpressed. Therefore, down-regulation of Otud-6b expression after prolonged cytokine stimulation may be required for cell proliferation in B lymphocytes.