MilliporeSigma
  • Dimerization of butenolide structures. A biomimetic approach to the dimeric sesquiterpene lactones (+/-)-biatractylolide and (+/-)-biepiasterolide.
Dimerization of butenolide structures. A biomimetic approach to the dimeric sesquiterpene lactones (+/-)-biatractylolide and (+/-)-biepiasterolide.
The Journal of organic chemistry (2004-12-22)
Sharanjeet K Bagal, Robert M Adlington, Jack E Baldwin, Rodolfo Marquez
ABSTRACT
The biomimetic synthesis of the bisesquiterpene lactones (+/-)-biatractylolide 1 and (+/-)-biepiasterolide 2 via dimerization of the captodative stabilized radical 8 is reported. Atractylon 7 has also been shown to be a possible intermediate during the biosynthesis of biatractylolide 1, biepiasterolide 2, atractylolide 3, and hydroxyatractylolide 4.
Sigma-Aldrich
Chlorotris(triphenylphosphine)cobalt(I), 97%