MilliporeSigma
  • Leptin augments coronary vasoconstriction and smooth muscle proliferation via a Rho-kinase-dependent pathway.
Leptin augments coronary vasoconstriction and smooth muscle proliferation via a Rho-kinase-dependent pathway.
Basic research in cardiology (2016-03-16)
Jillian N Noblet, Adam G Goodwill, Daniel J Sassoon, Alexander M Kiel, Johnathan D Tune
ABSTRACT
Leptin has been implicated as a key upstream mediator of pathways associated with coronary vascular dysfunction and disease. The purpose of this investigation was to test the hypothesis that leptin modifies the coronary artery proteome and promotes increases in coronary smooth muscle contraction and proliferation via influences on Rho kinase signaling. Global proteomic assessment of coronary arteries from lean swine cultured with obese concentrations of leptin (30 ng/mL) for 3 days revealed significant alterations in the coronary artery proteome (68 proteins) and identified an association between leptin treatment and calcium signaling/contraction (four proteins) and cellular growth and proliferation (35 proteins). Isometric tension studies demonstrated that both acute (30 min) and chronic (3 days, serum-free media) exposure to obese concentrations of leptin potentiated depolarization-induced contraction of coronary arteries. Inhibition of Rho kinase significantly reduced leptin-mediated increases in coronary artery contractions. The effects of leptin on the functional expression of Rho kinase were time-dependent, as acute treatment increased Rho kinase activity while chronic (3 day) exposure was associated with increases in Rho kinase protein abundance. Proliferation assays following chronic leptin administration (8 day, serum-containing media) demonstrated that leptin augmented coronary vascular smooth muscle proliferation and increased Rho kinase activity. Inhibition of Rho kinase significantly reduced these effects of leptin. Taken together, these findings demonstrate that leptin promotes increases in coronary vasoconstriction and smooth muscle proliferation and indicate that these phenotypic effects are associated with alterations in the coronary artery proteome and dynamic effects on the Rho kinase pathway.
Sigma-Aldrich
Bradykinin acetate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
Rho-associated Kinase (ROCK) Activity Assay, This Rho-associated Protein Kinase (ROCK) Activity Assay Kit is an enzyme immunoassay for detection of the active ROCK & DMPK family kinases.
Roche
5-Bromo-2′-deoxy-uridine Labeling and Detection Kit II, sufficient for ≤100 tests, storage temp.:-20°C