MilliporeSigma
MicroRNA-192* impairs adipocyte triglyceride storage.
Biochimica et biophysica acta (2016-01-10)
Raghavendra Mysore, You Zhou, Sanja Sädevirta, Hanna Savolainen-Peltonen, P A Nidhina Haridas, Jarkko Soronen, Marja Leivonen, Antti-Pekka Sarin, Pamela Fischer-Posovszky, Martin Wabitsch, Hannele Yki-Järvinen, Vesa M Olkkonen
ABSTRACT
We investigated the expression of miR-192* (miR-192-3p) in the visceral adipose tissue (VAT) of obese subjects and its function in cultured human adipocytes. This miRNA is a 3' arm derived from the same pre-miRNA as miR-192 (miR-192-5p) implicated in type 2 diabetes, liver disease and cancers, and is predicted to target key genes in lipid metabolism. In morbidly obese subjects undergoing bariatric surgery preceded by a very low calorie diet, miR-192* in VAT correlated negatively (r=-0.387; p=0.046) with serum triglyceride (TG) and positively with high-density lipoprotein (HDL) concentration (r=0.396; p=0.041). In a less obese patient cohort, the miRNA correlated negatively with the body mass index (r=-0.537; p=0.026). To characterize the function of miR-192*, we overexpressed it in cultured adipocytes and analyzed the expression of adipogenic differentiation markers as well as cellular TG content. Reduced TG and expression of the adipocyte marker proteins aP2 (adipocyte protein 2) and perilipin 1 were observed. The function of miR-192* was further investigated by transcriptomic profiling of adipocytes expressing this miRNA, revealing impacts on key lipogenic genes. A number of the mRNA alterations were validated by qPCR. Western analysis confirmed a marked reduction of the lipogenic enzyme SCD (stearoyl coenzyme A desaturase-1), the fatty aldehyde dehydrogenase ALDH3A2 (aldehyde dehydrogenase 3 family member A2) and the high-density lipoprotein receptor SCARB1 (scavenger receptor B, type I). SCD and ALDH3A2 were demonstrated to be direct targets of miR-192*. To conclude, the present data identify miR-192* as a novel controller of adipocyte differentiation and lipid homeostasis.
Sigma-Aldrich
Anti-ALDH3A2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FABP4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution