MilliporeSigma
  • An extremely efficient three-component reaction of aldehydes/ketones, amines, and phosphites (Kabachnik-Fields reaction) for the synthesis of alpha-aminophosphonates catalyzed by magnesium perchlorate.
An extremely efficient three-component reaction of aldehydes/ketones, amines, and phosphites (Kabachnik-Fields reaction) for the synthesis of alpha-aminophosphonates catalyzed by magnesium perchlorate.
The Journal of organic chemistry (2007-01-27)
Srikant Bhagat, Asit K Chakraborti
ABSTRACT
Commercially available magnesium perchlorate is reported as an extremely efficient catalyst for the synthesis of alpha-aminophosphonates. A three-component reaction (3-CR) of an amine, an aldehyde or a ketone, and a di-/trialkyl phosphite (Kabachnik-Fields reaction) took place in one pot under solvent-free conditions to afford the corresponding alpha-aminophosphonates in high yields and short times. The use of solvent retards the rate of the reaction and requires a much longer reaction time than that for neat conditions. The reactions involving an aldehyde, an aromatic amine without any electron-withdrawing substituent, and a phosphite are carried out at rt. The reactions involving cyclic ketones, aromatic amines with an electron-withdrawing substituent, and aryl alkyl ketone (e.g., acetophenone) require longer reaction times at rt or heating. Magnesium perchlorate was found to be superior to other metal perchlorates and metal triflates during the reaction of 4-methoxybenzaldehyde, 2,4-dinitroaniline, and dimethyl phosphite. The catalytic activity of various magnesium compounds was influenced by the counteranion, and magnesium perchlorate was found to be the most effective. The reaction was found to be general with di-/trialkyl phosphites and diaryl phosphite. The Mg(ClO4)2-catalyzed alpha-aminophosphonate synthesis in the present study perhaps represents a true three-component reaction as no intermediate formation of either an imine or alpha-hydroxy phosphonate was observed that indicated the simultaneous involvement of the carbonyl compound, the amine, and the phosphite in the transition state.
Sigma-Aldrich
Magnesium perchlorate, puriss. p.a., drying agent, ACS reagent, ≥98.0% (calc. based on dry substance, KT)
Sigma-Aldrich
Magnesium perchlorate, ACS reagent
Sigma-Aldrich
Magnesium perchlorate, puriss., free-flowing powder, ≥99.0% (calc. based on dry substance, KT)
Sigma-Aldrich
Aminoacetaldehyde dimethyl acetal, 99%