MilliporeSigma
  • EPAS1/HIF-2 alpha-mediated downregulation of tissue factor pathway inhibitor leads to a pro-thrombotic potential in endothelial cells.
EPAS1/HIF-2 alpha-mediated downregulation of tissue factor pathway inhibitor leads to a pro-thrombotic potential in endothelial cells.
Biochimica et biophysica acta (2016-01-31)
Benedicte Stavik, Sandra Espada, Xue Yan Cui, Nina Iversen, Sverre Holm, Marie-Christine Mowinkel, Bente Halvorsen, Grethe Skretting, Per Morten Sandset
ABSTRACT
Neovascularization and hemorrhaging are evident in advanced atherosclerotic plaques due to hypoxic conditions, and mediate the accumulation of metabolic substrates, inflammatory cells, lipids, and other blood born factors inside the plaque. Tissue factor (TF) pathway inhibitor (TFPI) is mainly expressed by endothelial cells and is the endogenous inhibitor of the coagulation activator TF, which together with the downstream product thrombin can drive plaque progression and atherogenesis. We aimed to investigate the effect of hypoxic conditions on endothelial cell expression and activity of TFPI and TF that are important in coagulation initiation. Hypoxia was induced in primary human umbilical vein endothelial cells using chemicals or 1% oxygen tension, and mRNA and protein expressions were measured using qRT-PCR, ELISA, and Western blot analysis. Microscopy of fluorescence-labeled cells was used to visualize cell-associated TFPI. Cell-surface factor Xa (FXa) activity was measured using a two-stage chromogenic substrate method. We found that hypoxia reduced the TFPI mRNA and protein levels and increased the TF mRNA expression in a dose-dependent manner. The effect on TFPI was apparent on all the protein pools of TFPI, i.e., secreted TFPI, cell-surface associated TFPI, and intracellular TFPI, and seemed to be dependent upon hypoxia inducible factor-2α (HIF-2α). An increase in FXa activity was also observed on the endothelial cell surface, reflecting an increase in pro-thrombotic potential of the cells. Our findings indicate that hypoxic conditions may enhance the pro-coagulant activity of endothelial cells, which may promote atherogenesis in addition to clinical events and thus the severity of atherosclerotic disorders.
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, case of 48 × 15 mL, histological tissue fixative
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Supelco
Zinc Standard for ICP, TraceCERT®, 1 g/L Zn in nitric acid (nominal concentration)
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
RIPA Buffer