MilliporeSigma
  • Molybdenum carbide as a highly selective deoxygenation catalyst for converting furfural to 2-methylfuran.
Molybdenum carbide as a highly selective deoxygenation catalyst for converting furfural to 2-methylfuran.
ChemSusChem (2014-04-24)
Ke Xiong, Wen-Sheng Lee, Aditya Bhan, Jingguang G Chen
ABSTRACT
Selectively cleaving the C=O bond outside the furan ring of furfural is crucial for converting this important biomass-derived molecule to value-added fuels such as 2-methylfuran. In this work, a combination of density functional theory (DFT) calculations, surface science studies, and reactor evaluation identified molybdenum carbide (Mo2 C) as a highly selective deoxygenation catalyst for converting furfural to 2-methylfuran. These results indicate the potential application of Mo2 C as an efficient catalyst for the selective deoxygenation of biomass-derived oxygenates including furanics and aromatics.
Sigma-Aldrich
2-Methylfuran, contains 200-400 BHT as stabilizer, 99%