MilliporeSigma
  • The mutation in Chd7 causes misexpression of Bmp4 and developmental defects in telencephalic midline.
The mutation in Chd7 causes misexpression of Bmp4 and developmental defects in telencephalic midline.
The American journal of pathology (2012-06-05)
Xuan Jiang, Yue Zhou, Li Xian, Weiqian Chen, Hanwei Wu, Xiang Gao
ABSTRACT
Mutations in chromosome-helicase-DNA-binding protein 7 (CHD7) are identified as the main cause for CHARGE syndrome (coloboma, heart anomaly, choanal atresia, retardation, genital and ear anomalies). Most patients (55% to 85%) with CHARGE syndrome display developmental defects in the central nervous system (CNS), of which pathology and molecular mechanisms remain unclear. In this study, we report a novel mutant mouse strain carrying a nonsense mutation, COA1, in exon4 of Chd7 gene. Chd7(COA1/+) mice phenocopied human CHARGE syndrome and displayed developmental defects in the telencephalic midline, including dilated third and lateral ventricles, reduced cerebral cortex, and corpus callosum crossing failure. Programed cell death in the telencephalic midline zone of Chd7(COA1/+) embryos was impaired, consistent with the incomplete telencephalic medial invagination in Chd7(COA1/+) embryos. Interestingly, expression of Bmp4, a signal well known to induce forebrain midline cell fate and apoptosis, was down-regulated and also expanded in the forebrain of Chd7(COA1/+) embryos. Furthermore, in vitro studies suggested that CHD7 may directly regulate Bmp4 expression by binding with an enhancer element downstream of the Bmp4 locus. These studies provide novel insight into pathogenesis of CNS anomalies in CHARGE syndrome.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Roche
SP6 RNA Polymerase, from Escherichia coli BL 21/pSR3
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution