MilliporeSigma
  • 3-Hydroxyisobutyrate dehydrogenase, an impurity in commercial 3-hydroxybutyrate dehydrogenase.
3-Hydroxyisobutyrate dehydrogenase, an impurity in commercial 3-hydroxybutyrate dehydrogenase.
The Biochemical journal (1987-01-01)
E B Worrall, S Gassain, D J Cox, M C Sugden, T N Palmer
ABSTRACT
The enzymic determination of D-3-hydroxybutyrate and acetoacetate normally involves the use of 3-hydroxybutyrate dehydrogenase (HBDH, EC 1.1.1.30) of bacterial origin. We show that HBDH from Rhodopseudomonas spheroides (BCL, grade II) contains a 3-hydroxyisobutyrate dehydrogenase (HIBDH) activity: activity with 3-hydroxyisobutyrate as substrate was greater than 10% of that with 3-hydroxybutyrate. However, HBDH could be prepared essentially free of HIBDH activity by incubation at 37 degrees C in the presence of 1 mM-CaCl2, to produce an enzyme preparation that may be used for the specific determination of 3-hydroxybutyrate. Use of the purified enzyme preparations indicated that a major product of valine metabolism in hemidiaphragms from 40 h-starved rats was 3-hydroxyisobutyrate rather than 3-hydroxybutyrate.
Product Description
Roche
3-Hydroxybutyrate Dehydrogenase (3-HBDH), suspension, ~3 units/mg protein (At 25 °C with 3-hydroxybutyrate as the substrate; standardized with BSA.), grade II