MilliporeSigma
  • IGFBP3 promotes esophageal cancer growth by suppressing oxidative stress in hypoxic tumor microenvironment.
IGFBP3 promotes esophageal cancer growth by suppressing oxidative stress in hypoxic tumor microenvironment.
American journal of cancer research (2014-02-01)
Mitsuteru Natsuizaka, Hideaki Kinugasa, Shingo Kagawa, Kelly A Whelan, Seiji Naganuma, Harry Subramanian, Sanders Chang, Kei J Nakagawa, Naryan L Rustgi, Yoshiaki Kita, Shoji Natsugoe, Devraj Basu, Phyllis A Gimotty, Andres J Klein-Szanto, J Alan Diehl, Hiroshi Nakagawa
ABSTRACT
Insulin-like growth factor binding protein 3 (IGFBP3), a hypoxia-inducible gene, regulates a variety of cellular processes including cell proliferation, senescence, apoptosis and epithelial-mesenchymal transition (EMT). IGFBP3 has been linked to the pathogenesis of cancers. Most previous studies focus upon proapoptotic tumor suppressor activities of IGFBP3. Nevertheless, IGFBP3 is overexpressed in certain cancers including esophageal squamous cell carcinoma (ESCC), one of the most aggressive forms of squamous cell carcinomas (SCCs). The tumor-promoting activities of IGFBP3 remain poorly understood in part due to a lack of understanding as to how the tumor microenvironment may influence IGFBP3 expression and how IGFBP3 may in turn influence heterogeneous intratumoral cell populations. Here, we show that IGFBP3 overexpression is associated with poor postsurgical prognosis in ESCC patients. In xenograft transplantation models with genetically engineered ESCC cells, IGFBP3 contributes to tumor progression with a concurrent induction of a subset of tumor cells showing high expression of CD44 (CD44H), a major cell surface receptor for hyaluronic acid, implicated in invasion, metastasis and drug resistance. Our gain-of-function and loss-of-function experiments reveal that IGFBP3 mediates the induction of intratumoral CD44H cells. IGFBP3 cooperates with hypoxia to mediate the induction of CD44H cells by suppressing reactive oxygen species (ROS) in an insulin-like growth factor-independent fashion. Thus, our study sheds light on the growth stimulatory functions of IGFPB3 in cancer, gaining a novel mechanistic insight into the functional interplay between the tumor microenvironment and IGFBP3.
Product Description
Roche
DNase I, grade II, from bovine pancreas