MilliporeSigma
  • Cloning and characterization of PRAX-1. A new protein that specifically interacts with the peripheral benzodiazepine receptor.
Cloning and characterization of PRAX-1. A new protein that specifically interacts with the peripheral benzodiazepine receptor.
The Journal of biological chemistry (1999-01-23)
S Galiègue, O Jbilo, T Combes, E Bribes, P Carayon, G Le Fur, P Casellas
ABSTRACT
Using a cytoplasmic domain of the peripheral benzodiazepine receptor (PBR) as a bait in the yeast two-hybrid system, we have isolated a cDNA encoding a new protein that specifically interacts with PBR. We named it PRAX-1, for peripheral benzodiazepine receptor-associated protein 1. PRAX-1 is a 1857-amino acid protein, the sequence of which was structurally unrelated to any known proteins. The gene encoding PRAX-1 is located in the q22-q23 region of the long arm of the human chromosome 17. The PRAX-1 mRNA is 7.5 kilobase pairs, predominantly expressed in the central nervous system, pituitary gland, and thymus. At the protein level, we found the PRAX-1 as a single 220-250-kDa protein in the brain and in many different human cell lines tested using specific antibody raised against PRAX-1. Parallel analysis of the PRAX-1 mRNA and protein expression performed in mouse and rat gave similar results. Immunocytochemistry analysis carried out to define the distribution of the PRAX-1 protein in the rat brain showed that PRAX-1 was prevalent in the mesolimbic system, specially abundant in the CA1 subfield of the hippocampus. Exhibiting several domains involved in protein-protein interaction (three proline-rich domains, three leucine-zipper motifs, and an Src homology region 3-like domain), the PRAX-1 may be looked upon as a new adaptator protein. We show that both the Src homology region 3-like domain and a proline-rich domain in PRAX-1 are required for the interaction with PBR. PRAX-1 is a cytoplasmic protein that also partially colocalizes with PBR in the mitochondria, as determined by confocal microscopy and Western blotting. Altogether our observations support a model of interaction implicating PBR and this newly described protein, PRAX-1. As being the first cytoplasmic protein associated with PBR, PRAX-1 is a new tool that opens new fields for exploring PBR biological roles.