MilliporeSigma
  • Synthesis and biological evaluation of 10,11-methylenedioxy-14-azacamptothecin.
Synthesis and biological evaluation of 10,11-methylenedioxy-14-azacamptothecin.
Organic letters (2006-07-28)
Mark A Elban, Wenyue Sun, Brian M Eisenhauer, Rong Gao, Sidney M Hecht
ABSTRACT
[reaction: see text] 10,11-Methylenedioxy-14-azacamptothecin, a potent analogue of the antitumor agent camptothecin (CPT), has been prepared via a key condensation between AB and DE ring precursors. The biological testing of this compound validated a strategy for modulation of the off-rate of camptothecin analogues from the topoisomerase-DNA-CPT ternary complex via structural modification.