MilliporeSigma
  • Ultrathin Printable Graphene Supercapacitors with AC Line-Filtering Performance.
Ultrathin Printable Graphene Supercapacitors with AC Line-Filtering Performance.
Advanced materials (Deerfield Beach, Fla.) (2015-05-15)
Zhong-Shuai Wu, Zhaoyang Liu, Khaled Parvez, Xinliang Feng, Klaus Müllen
ABSTRACT
Ultrathin printable graphene supercapacitors are demonstrated, based on solution-processed electrochemically exfoliated graphene hybrid films on an ultrathin poly(ethylene terephthalate) substrate, exhibiting an unprecedented volumetric capacitance of 348 F cm(-3) , an ultrahigh scan rate of 2000 V s(-1) , and AC line-filtering performance.
Sigma-Aldrich
Graphene/PEDOT:PSS hybrid ink, dispersion in DMF, avg. no. of layers, 1 ‑ 3
Sigma-Aldrich
Manganese Oxide (MnO2) nanosheets