MilliporeSigma
  • Investigating physical chromatin associations across the Xenopus genome by chromatin immunoprecipitation.
Investigating physical chromatin associations across the Xenopus genome by chromatin immunoprecipitation.
Cold Spring Harbor protocols (2014-05-03)
George E Gentsch, James C Smith
ABSTRACT
Chromatin immunoprecipitation (ChIP) combined with genomic analysis techniques provide a global snapshot of protein-DNA interactions in the context of chromatin, yielding insights into which genomic loci might be regulated by the DNA-associated protein under investigation. This protocol describes how to perform ChIP on intact or dissected Xenopus embryos. The ChIP-isolated DNA fragments are suitable for high-throughput sequencing (ChIP-Seq) or for quantitative PCR (ChIP-qPCR). In this protocol, embryonic tissue is harvested from Xenopus tropicalis or Xenopus laevis at the developmental stage of interest, and DNA-associated proteins are immobilized to their endogenous genomic binding sites with formaldehyde. Nuclei are extracted from embryos and subjected to sonication so as to shear the chromatin to a size that allows sufficient positional resolution of protein binding to genomic DNA. Chromatin fragments bound by the protein of interest are immunoprecipitated using antibody-coupled beads, washed under high-stringency conditions, and stripped from the beads with anionic detergents. The chemical cross-links are reversed, and the coimmunoprecipitated DNA is purified. The resulting DNA fragments can be analyzed by qPCR or used to create a ChIP-Seq library. General advice for qPCR and for making ChIP-Seq libraries is offered, and approaches for analyzing ChIP-Seq data are outlined.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Roche
cOmplete ULTRA Tablets, EDTA-free, glass vials Protease Inhibitor Cocktail, Tablets provided in glass vials
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial