MilliporeSigma
  • Visible light-promoted metal-free C-H activation: diarylketone-catalyzed selective benzylic mono- and difluorination.
Visible light-promoted metal-free C-H activation: diarylketone-catalyzed selective benzylic mono- and difluorination.
Journal of the American Chemical Society (2013-11-05)
Ji-Bao Xia, Chen Zhu, Chuo Chen
ABSTRACT
We report herein an operationally simple method for the direct conversion of benzylic C-H groups to C-F. We show that visible light can activate diarylketones to abstract a benzylic hydrogen atom selectively. Adding a fluorine radical donor yields the benzylic fluoride and regenerates the catalyst. The selective formation of mono- and difluorination products can be achieved by catalyst control. 9-Fluorenone catalyzes benzylic C-H monofluorination, while xanthone catalyzes benzylic C-H difluorination. The scope and efficiency of this new C-H fluorination method are significantly better than those of the existing methods. This is also the first report of selective C-H gem-difluorination.