MilliporeSigma
  • Determination of the acaricide fenbutatin oxide in water samples by automated headspace-SPME-GC/MS.
Determination of the acaricide fenbutatin oxide in water samples by automated headspace-SPME-GC/MS.
Journal of separation science (2005-05-26)
Christophe Devos, Luc Moens, Pat Sandra
ABSTRACT
The analysis of the acaricide fenbutatin oxide (FBTO) having a molecular weight of 1052.66 g mol(-1) in water samples by capillary GC/MS after in-situ derivatization with sodium tetraethylborate (NaBEt4) and headspace-SPME enrichment is described. Automated SPME is performed at 80 degrees C for 30 min. Detection is carried out in the ion monitoring mode with deuterated triphenyltin (TPhTd15) as internal standard. Good linearity (R2 = 0.9993) was obtained in the dynamic range 20 to 1000 ng L(-1) with a limit of detection of 16 ng L(1) (LOD at 3 S/N) and a limit of quantitation of 50 ng L(-1) (LOQ at 10 S/N). Intra-day RSD% for n=6 was 8.9 at the LOQ level.
Product Description
Supelco
Fenbutatin oxide, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium tetraethylborate, 97%