MilliporeSigma
  • Synthesis of imidazoles through the copper-catalyzed cross-cycloaddition between two different isocyanides.
Synthesis of imidazoles through the copper-catalyzed cross-cycloaddition between two different isocyanides.
Journal of the American Chemical Society (2006-08-17)
Chikashi Kanazawa, Shin Kamijo, Yoshinori Yamamoto
ABSTRACT
The copper-catalyzed reaction between two different isocyanides produces the corresponding heteroaromatization products, imidazoles, in good yields. The reaction proceeds most probably through the activation of the sp3 C-H bond in the isocyanides by a copper catalyst, followed by a [3 + 2] cycloaddition between the cuprioisocyanide intermediates and arylisocyanides.
Sigma-Aldrich
Ethyl isocyanoacetate, 95%