MilliporeSigma
  • Palladium-catalyzed cross-coupling reactions of (2-pyridyl)allyldimethylsilanes with aryl iodides.
Palladium-catalyzed cross-coupling reactions of (2-pyridyl)allyldimethylsilanes with aryl iodides.
Organic letters (2006-02-14)
Toshiki Nokami, Yutaka Tomida, Toshiyuki Kamei, Kenichiro Itami, Jun-ichi Yoshida
ABSTRACT
[reaction: see text] (2-Pyridyl)allyldimethylsilanes were found to be novel pyridyl transfer reagents in palladium-catalyzed cross-coupling reactions of various aryl iodides in the presence of silver(I) oxide as an activator.
Sigma-Aldrich
2-(Allyldimethylsilyl)pyridine, 90%
Pricing and availability is not currently available.