MilliporeSigma
  • c-Cbl reduces stability of rescued ∆F508-CFTR in human airway epithelial cells: Implications for cystic fibrosis treatment.
c-Cbl reduces stability of rescued ∆F508-CFTR in human airway epithelial cells: Implications for cystic fibrosis treatment.
Communicative & integrative biology (2013-06-12)
Kristine M Cihil, Andrew Zimnik, Agnieszka Swiatecka-Urban
ABSTRACT
CFTR is a PKA activated Cl(-) channel expressed in the apical membrane of fluid transporting epithelia. We previously demonstrated that c-Cbl decreases CFTR stability in the plasma membrane by facilitating its endocytosis and lysosomal degradation in human airway epithelium. The most common mutation associated with cystic fibrosis, deletion of Phe508 (∆F508), leads to a temperature sensitive biosynthetic processing defect in the CFTR protein. Mature ∆F508-CFTR that has been rescued by low temperature or chemical chaperones is partially functional as a Cl(-) channel but has decreased plasma membrane stability due to altered post-maturational trafficking. Our present data demonstrate that c-Cbl controls the post-maturational trafficking of rescued ∆F508-CFTR. Partial depletion of c-Cbl increased stability of the plasma membrane associated mature ∆F508-CFTR and the ∆F508-CFTR mediated Cl(-) secretion. These data indicate that correcting the post-maturational trafficking of ∆F508-CFTR may represent a therapeutic approach complementary to the biosynthetic rescue. Because c-Cbl functions as an adaptor and scaffolding protein during CFTR endocytosis, we propose that interfering with the c-Cbl mediated endocytic recruitment of ∆F508-CFTR may increase stability of ∆508-CFTR in the plasma membrane after its biosynthetic rescue.