MilliporeSigma
  • HSCARG negatively regulates the cellular antiviral RIG-I like receptor signaling pathway by inhibiting TRAF3 ubiquitination via recruiting OTUB1.
HSCARG negatively regulates the cellular antiviral RIG-I like receptor signaling pathway by inhibiting TRAF3 ubiquitination via recruiting OTUB1.
PLoS pathogens (2014-04-26)
Yanyan Peng, Ruidan Xu, Xiaofeng Zheng
ABSTRACT
RIG-I like receptors (RLRs) recognize cytosolic viral RNA and initiate innate immunity; they increase the production of type I interferon (IFN) and the transcription of a series of antiviral genes to protect the host organism. Accurate regulation of the RLR pathway is important for avoiding tissue injury induced by excessive immune response. HSCARG is a newly reported negative regulator of NF-κB. Here we demonstrated that HSCARG participates in innate immunity. HSCARG inhibited the cellular antiviral response in an NF-κB independent manner, whereas deficiency of HSCARG had an opposite effect. After viral infection, HSCARG interacted with tumor necrosis receptor-associated factor 3 (TRAF3) and inhibited its ubiquitination by promoting the recruitment of OTUB1 to TRAF3. Knockout of HSCARG attenuated the de-ubiquitination of TRAF3 by OTUB1, and knockdown of OTUB1 abolished the effect of HSCARG. HSCARG also interacted with Ikappa-B kinase epsilon (IKKε) after viral infection and impaired the association between TRAF3 and IKKε, which further decreased the phosphorylation of IKKε and interferon response factor 3 (IRF3), thus suppressed the dimerization and nuclear translocation of IRF3. Moreover, knockdown of TRAF3 dampened the inhibitory effect of IFN-β transcription by HSCARG, suggesting that TRAF3 is necessary for HSCARG to down-regulate RLR pathway. This study demonstrated that HSCARG is a negative regulator that enables balanced antiviral innate immunity.
Sigma-Aldrich
MISSION® esiRNA, targeting human TRAF3
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid