MilliporeSigma
  • Regulation of gene expression in Neurospora crassa with a copper responsive promoter.
Regulation of gene expression in Neurospora crassa with a copper responsive promoter.
G3 (Bethesda, Md.) (2013-10-22)
Teresa M Lamb, Justin Vickery, Deborah Bell-Pedersen
ABSTRACT
Precise control of gene expression is a powerful method to elucidate biological function, and protein overexpression is an important tool for industry and biochemistry. Expression of the Neurospora crassa tcu-1 gene (NCU00830), encoding a high-affinity copper transporter, is tightly controlled by copper availability. Excess copper represses, and copper depletion, via the use of a copper chelator, activates expression. The kinetics of induction and repression of tcu-1 are rapid, and the effects are long lived. We constructed a plasmid carrying the bar gene (for glufosinate selection) fused to the tcu-1 promoter. This plasmid permits the generation of DNA fragments that can direct integration of Ptcu-1 into any desired locus. We use this strategy to integrate Ptcu-1 in front of wc-1, a circadian oscillator and photoreceptor gene. The addition of excess copper to the Ptcu-1::wc-1 strain phenocopies a Δwc-1 strain, and the addition of the copper chelator, bathocuproinedisulfonic acid, phenocopies a wc-1 overexpression strain. To test whether copper repression can recapitulate the loss of viability that an essential gene knockout causes, we placed Ptcu-1 upstream of the essential gene, hpt-1. The addition of excess copper drastically reduced the growth rate as expected. Thus, this strategy will be useful to probe the biological function of any N. crassa gene through controlled expression.
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Sigma-Aldrich
Bathocuproinedisulfonic acid disodium salt, for spectrophotometric det. of Cu, Fe
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis