MilliporeSigma
  • Palladium-catalyzed alkylation of sp2 and sp3 C-H bonds with methylboroxine and alkylboronic acids: two distinct C-H activation pathways.
Palladium-catalyzed alkylation of sp2 and sp3 C-H bonds with methylboroxine and alkylboronic acids: two distinct C-H activation pathways.
Journal of the American Chemical Society (2006-09-28)
Xiao Chen, Charles E Goodhue, Jin-Quan Yu
ABSTRACT
Palladium-catalyzed alkylations of sp2 and sp3 C-H bonds with either methylboroxine or alkylboronic acids were developed. Ag2O or AgCO3 is used as a crucial oxidant and promoter for the transmetalation step. Ether, ester, alcohol, and alkene functional groups are tolerated. A new C-H activation pathway differing from the cyclometalation process is elucidated using methylboroxine as the coupling partner.
Sigma-Aldrich
Cyclopropylboronic acid