MilliporeSigma
  • Angiotensin I converting enzyme inhibitory peptides obtained after in vitro hydrolysis of pea (Pisum sativum var. Bajka) globulins.
Angiotensin I converting enzyme inhibitory peptides obtained after in vitro hydrolysis of pea (Pisum sativum var. Bajka) globulins.
BioMed research international (2014-09-25)
Anna Jakubczyk, Barbara Baraniak
ABSTRACT
Pea seeds represent a valuable source of active compounds that may positively influence health. In this study, the pea globulins were digested in vitro under gastrointestinal condition and potentially bioaccessible angiotensin I converting enzyme (ACE) inhibitory peptides were identified. The degree of hydrolysis after pepsin, 14.42%, and pancreatin, 30.65%, were noted. The peptides with the highest ACE inhibitory properties were separated using ion exchange chromatography on DEAE-cellulose. Thirteen peptides fractions were obtained but only four showed potential antihypertensive properties. The highest inhibitory activity was determined for the fraction F8 (IC50 = 0.0014 mg/mL). This fraction was separated on Sephadex G10 and two peptide fractions were obtained. The peptides fraction (B) with the highest ACE inhibitory activity (IC50 = 0.073 mg/mL) was identified by ESI-MS/MS. The sequences of ACE inhibitory peptides were GGSGNY, DLKLP, GSSDNR, MRDLK, and HNTPSR. Based on Lineweaver-Burk plots for the fraction B, the kinetic parameters as K m, Vmax, and K i and mode of inhibition were determined. This fraction belongs to uncompetitive inhibitor of ACE activity. The seeds of pea are the source of precursor protein, which releases the ACE inhibitory peptides as a result of enzymatic hydrolysis.
Sigma-Aldrich
α-Amylase from hog pancreas, powder, ~50 U/mg