MilliporeSigma
  • A growth-rate composition formula for the growth of E.coli on co-utilized carbon substrates.
A growth-rate composition formula for the growth of E.coli on co-utilized carbon substrates.
Molecular systems biology (2015-04-12)
Rutger Hermsen, Hiroyuki Okano, Conghui You, Nicole Werner, Terence Hwa
ABSTRACT
When bacteria are cultured in medium with multiple carbon substrates, they frequently consume these substrates simultaneously. Building on recent advances in the understanding of metabolic coordination exhibited by Escherichia coli cells through cAMP-Crp signaling, we show that this signaling system responds to the total carbon-uptake flux when substrates are co-utilized and derive a mathematical formula that accurately predicts the resulting growth rate, based only on the growth rates on individual substrates.
Product Description
Supelco
Glucose (HK) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
Free Glycerol Determination Kit, 1 kit sufficient for 1000 reactions