MilliporeSigma
  • A chemical-genetic interaction map of small molecules using high-throughput imaging in cancer cells.
A chemical-genetic interaction map of small molecules using high-throughput imaging in cancer cells.
Molecular systems biology (2015-12-25)
Marco Breinig, Felix A Klein, Wolfgang Huber, Michael Boutros
ABSTRACT
Small molecules often affect multiple targets, elicit off-target effects, and induce genotype-specific responses. Chemical genetics, the mapping of the genotype dependence of a small molecule's effects across a broad spectrum of phenotypes can identify novel mechanisms of action. It can also reveal unanticipated effects and could thereby reduce high attrition rates of small molecule development pipelines. Here, we used high-content screening and image analysis to measure effects of 1,280 pharmacologically active compounds on complex phenotypes in isogenic cancer cell lines which harbor activating or inactivating mutations in key oncogenic signaling pathways. Using multiparametric chemical-genetic interaction analysis, we observed phenotypic gene-drug interactions for more than 193 compounds, with many affecting phenotypes other than cell growth. We created a resource termed the Pharmacogenetic Phenome Compendium (PGPC), which enables exploration of drug mode of action, detection of potential off-target effects, and the generation of hypotheses on drug combinations and synergism. For example, we demonstrate that MEK inhibitors amplify the viability effect of the clinically used anti-alcoholism drug disulfiram and show that the EGFR inhibitor tyrphostin AG555 has off-target activity on the proteasome. Taken together, this study demonstrates how combining multiparametric phenotyping in different genetic backgrounds can be used to predict additional mechanisms of action and to reposition clinically used drugs.
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Caffeic acid phenethyl ester, ≥97% (HPLC), powder
Sigma-Aldrich
Z-L-Phe chloromethyl ketone, 98%
Sigma-Aldrich
DMAT, ≥98% (HPLC)
Sigma-Aldrich
DAPH, ≥98% (HPLC), solid
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%