MilliporeSigma
Efficient access to cyclic ureas via Pd-catalyzed cyclization.
Organic letters (2006-07-14)
Mark McLaughlin, Michael Palucki, Ian W Davies
ABSTRACT
[Structure: see text] An efficient regioselective method for the preparation of structurally diverse imidazopyridinones and benzoimidazolones starting from readily available and economical starting materials is described. High-yielding reductive alkylation of electron-deficient o-haloarylamines followed by treatment with inexpensive N-chlorosulfonyl isocyanate afforded primary ureas in good overall yields. A Pd-catalyzed urea cyclization reaction furnished imidazopyridinones and benzoimidazolones in excellent yields. Overall, the developed chemistry provides rapid access to pharmaceutically important heterocyclic compounds with high efficiency.
Sigma-Aldrich
Palladium(II) acetate, reagent grade, 98%
Sigma-Aldrich
Palladium(II) Acetate ChemBeads