MilliporeSigma
  • Preventive and therapeutic potential of p38 alpha-selective mitogen-activated protein kinase inhibitor in nonobese diabetic mice with type 1 diabetes.
Preventive and therapeutic potential of p38 alpha-selective mitogen-activated protein kinase inhibitor in nonobese diabetic mice with type 1 diabetes.
The Journal of pharmacology and experimental therapeutics (2006-04-11)
Satyanarayana Medicherla, Andrew A Protter, Jing Ying Ma, Ruban Mangadu, Ramona Almirez, Bruce Koppelman, Irene Kerr, Tony A Navas, Fabiola Movius, Mamatha Reddy, Yu-Wang Liu, Gregory Luedtke, John Perumattam, Babu Mavunkel, Sundeep Dugar, George F Schreiner
ABSTRACT
Mitogen-activated protein kinases (MAPKs) and heat shock proteins (HSPs) are ubiquitous proteins that function within T cells in both normal and stress-related pathophysiological states, including type 1 diabetes. The nonobese diabetic (NOD) mouse spontaneously develops T cell-mediated autoimmune pancreatic beta cell destruction that is similar to type 1 diabetes in humans. Because p38 MAPKs have been shown to modulate T cell function, we studied the effects of a p38alpha MAPK-selective inhibitor, indole-5-carboxamide (SD-169), on the development and progression of type 1 diabetes in the NOD mouse. In preventive treatment studies, SD-169 significantly reduced p38 and HSP60 expression in T cells of the pancreatic beta islets. Following treatment, the incidence of diabetes as determined by blood glucose levels was significantly lower, and immuno-histochemistry of pancreatic beta islet tissue demonstrated significant reduction in CD5+ T cell infiltration in the SD-169 treatment group as compared with untreated NOD mice. In therapeutic studies using mildly and moderately hyperglycemic NOD mice, SD-169 treatment lowered blood glucose and improved glucose homeostasis. Furthermore, following cessation of SD-169 treatment, NOD mice showed significant arrest of diabetes. In conclusion, we report that this p38alpha-selective inhibitor prevents the development and progression of diabetes in NOD mice by inhibiting T cell infiltration and activation, thereby preserving beta cell mass via inhibition of the p38 MAPK signaling pathway. These results have bearing on current prophylactic and therapeutic protocols using p38alpha-selective inhibitors in the prediabetic period for children at high risk of type 1 diabetes, in the honeymoon period, and for adults with latent autoimmune diabetes.
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, biotin-SP conjugate, Species Adsorbed, 1 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, biotin-SP conjugate, Species Adsorbed, 1.5 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, biotin-SP conjugate, Species Adsorbed, 1.1 mg/mL, Chemicon®
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, biotin conjugate, Species Adsorbed, 0.8 mg/mL (after reconstitution), Chemicon®