- A concise asymmetric synthesis of (2S,3S,7S)-3,7-dimethylpentadecan-2-yl Acetate and propionate, the sex pheromones of pine sawflies.
A concise asymmetric synthesis of (2S,3S,7S)-3,7-dimethylpentadecan-2-yl Acetate and propionate, the sex pheromones of pine sawflies.
The Journal of organic chemistry (2004-05-22)
Pei-Qiang Huang, Hong-Qiao Lan, Xiao Zheng, Yuan-Ping Ruan
PMID15153034
ABSTRACT
(2S,3S,7S)-3,7-Dimethylpentadecan-2-yl acetate (2) and its propionate analogue (3) are the main sex pheromones of all Neodiprion species and Diprion similes, respectively. Starting from (S)-malic acid and employing a highly chemo-, regio-, and stereoselective tandem ester reduction-epoxide formation-reductive epoxide-opening reaction protocol, an efficient total synthesis of (2S,3S,7S)-2 and -3 is reported herein.