MilliporeSigma
  • Heteropoly acid as a novel nitrene transfer agent: a facile and practical aziridination of olefins with Chloramine-T.
Heteropoly acid as a novel nitrene transfer agent: a facile and practical aziridination of olefins with Chloramine-T.
Chemical communications (Cambridge, England) (2004-04-08)
G D Kishore Kumar, Sundarababu Baskaran
ABSTRACT
Environmentally benign HPA is found to be an efficient catalyst for aziridination of olefins in the presence of inexpensive Chloramine-T as a nitrogen source: instantaneous at room temperature, requires only stoichiometric amount of olefin and no allyl amine side product.
Sigma-Aldrich
Chloramine-T hydrate, 95%
Sigma-Aldrich
Chloramine T trihydrate, ACS reagent, 98%