MilliporeSigma
  • Selective oxidation of glycosyl sulfides to sulfoxides using magnesium monoperoxyphthalate and microwave irradiation.
Selective oxidation of glycosyl sulfides to sulfoxides using magnesium monoperoxyphthalate and microwave irradiation.
The Journal of organic chemistry (2004-04-13)
Ming-Yi Chen, Laxmikant Narhari Patkar, Chun-Cheng Lin
ABSTRACT
A protocol that uses moist magnesium monoperoxyphthalate (MMPP) as an oxidant under microwave irradiation rapidly yields a variety of glycosyl sulfoxides from corresponding sulfides in high yields with high selectivity.
Sigma-Aldrich
Magnesium bis(monoperoxyphthalate) hexahydrate, 80%, technical grade