MilliporeSigma
  • Temporal control of a dendritogenesis-linked gene via REST-dependent regulation of nuclear factor I occupancy.
Temporal control of a dendritogenesis-linked gene via REST-dependent regulation of nuclear factor I occupancy.
Molecular biology of the cell (2011-01-29)
Wei Wang, Yong Shin, Min Shi, Daniel L Kilpatrick
ABSTRACT
Developing neurons undergo a series of maturational stages, and the timing of these events is critical for formation of synaptic circuitry. Here we addressed temporal regulation of the Gabra6 gene, which is expressed in a delayed manner during dendritogenesis in maturing cerebellar granule neurons (CGNs). Developmental up-regulation of Gabra6 transcription required a binding site for nuclear factor I (NFI) proteins. The amounts and DNA binding activities of NFI proteins were similar in immature and mature CGNs; however, NFI occupancy of the Gabra6 promoter in native chromatin was temporally delayed in parallel with Gabra6 gene expression, both in vivo and in culture. The trans-repressor RE1 silencing transcription factor (REST) occupied the Gabra6 proximal promoter in CGN progenitors and early postmitotic CGNs, and its departure mirrored the initial onset of NFI binding as CGNs differentiated. Furthermore constitutive REST expression blocked both Gabra6 expression and NFI occupancy in mature CGNs, whereas REST knockdown in immature CGNs accelerated the initiation of both events. These studies identify a novel mechanism for controlling the timing of dendritogenesis-associated gene expression in maturing neurons through delayed binding of NFI proteins to chromatin. They also establish a temporal function for REST in preventing premature promoter occupancy by NFI proteins in early-stage postmitotic neurons.
Sigma-Aldrich
MISSION® esiRNA, targeting human REST