MilliporeSigma
  • Discovery of a potent, orally available, and isoform-selective retinoic acid beta2 receptor agonist.
Discovery of a potent, orally available, and isoform-selective retinoic acid beta2 receptor agonist.
Journal of medicinal chemistry (2005-11-24)
Birgitte W Lund, Fabrice Piu, Natalie K Gauthier, Anne Eeg, Erika Currier, Vladimir Sherbukhin, Mark R Brann, Uli Hacksell, Roger Olsson
ABSTRACT
4'-Octyl-4-biphenylcarboxylic acid (1g, AC-55649) was identified as a highly isoform-selective agonist at the human RARbeta2 receptor in a functional intact cell-based screening assay. The subsequent hit to lead optimization transformed the lipophilic, poorly soluble hit into a more potent and orally available compound (2, AC-261066) with retained beta2 selectivity and greatly improved physiochemical properties. Being an isoform-selective RARbeta2 receptor agonist that discriminates between nuclear receptor isoforms having identical ligand binding domains, 2 will be useful as a pharmacological research tool but also a valuable starting point for drug development.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
AC261066, ≥98% (HPLC)