MilliporeSigma
  • POSaM: a fast, flexible, open-source, inkjet oligonucleotide synthesizer and microarrayer.
POSaM: a fast, flexible, open-source, inkjet oligonucleotide synthesizer and microarrayer.
Genome biology (2004-08-04)
Christopher Lausted, Timothy Dahl, Charles Warren, Kimberly King, Kimberly Smith, Michael Johnson, Ramsey Saleem, John Aitchison, Lee Hood, Stephen R Lasky
ABSTRACT
DNA arrays are valuable tools in molecular biology laboratories. Their rapid acceptance was aided by the release of plans for a pin-spotting microarrayer by researchers at Stanford. Inkjet microarraying is a flexible, complementary technique that allows the synthesis of arrays of any oligonucleotide sequences de novo. We describe here an open-source inkjet arrayer capable of rapidly producing sets of unique 9,800-feature arrays.
Sigma-Aldrich
3-Methoxypropionitrile, ≥98.0% (GC)