MilliporeSigma
The cytoskeletal protein Ndel1 regulates dynamin 2 GTPase activity.
PloS one (2011-02-02)
Mathieu Chansard, Jian Wang, Hong Chi Tran, Gernot Neumayer, Su Yeon Shim, Young-Un Park, Camille Belzil, Hoa Thi Le, Sang Ki Park, Minh Dang Nguyen
ABSTRACT
Cytoskeleton dynamics, membranes trafficking and positioning are essential for the proper functioning of any mammalian cell. The identification of the molecules and mechanisms that allow these cellular processes to interface is vital for understanding cell behaviors. Ndel1, the mammalian homolog of the Aspergillus nidulans NudE, organizes the cytoskeleton and regulates molecular motors, thereby impacting on the positioning of membranes. Hypothetically, Ndel1 can act in concert with enzymes controlling membrane trafficking (vesicle-mediated transport) per se, but this idea has never been investigated. We now report that a pool of Ndel1 associates directly with Dynamin 2 (Dyn2), a large cytosolic GTPase involved in the trafficking of the AMPA receptor subunit GluR1. In vitro, Ndel1 enhances Dyn2 GTPase activity in its unassembled and assembled forms, without promoting oligomerization of the enzyme. In cells, gain and loss of function of Ndel1 recapitulate the effects of overexpression of Dyn2 and Dyn2 dominant negative with reduced GTPase activity on the intracellular localization of GluR1, respectively, without affecting the stability of microtubules. Together, these results indicate that Ndel1 regulates Dyn2 GTPase activity and impacts GluR1-containing membranes distribution in a manner reminiscent of Dyn2.
Sigma-Aldrich
MISSION® esiRNA, targeting human NDEL1
Sigma-Aldrich
Endoplasmic Reticulum Isolation Kit, isolation of intact ER from mammalian soft tissues and cultured cells