MilliporeSigma
  • Electrophilic fluorinating reagent mediated synthesis of fluorinated alpha-keto Ethers, benzil, and 6,6'-dialkoxy-2,2'-bipyridines.
Electrophilic fluorinating reagent mediated synthesis of fluorinated alpha-keto Ethers, benzil, and 6,6'-dialkoxy-2,2'-bipyridines.
The Journal of organic chemistry (2002-08-31)
Sudha Manandhar, Rajendra P Singh, Gary V Eggers, Jean'ne M Shreeve
ABSTRACT
Interactions of various fluorinated and nonfluorinated alcohols with trans-stilbene in the presence of electrophilic reagents were studied. Under neat conditions, reactions of trans-stilbene (1) with fluorinated alcohols, R(f)OH (R(f) = CF3CH2-, CFH2CH2-, CF3CF2CH2-, CF2H(CF2)3CH2-, (CF3)2CH-, (CF3)3C- (2a-f) in the presence of an electrophilic reagent, 1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) (Selectfluor) or N,N-difluoro-2,2'-bipyridinium bis(tetrafluoroborate) (MEC-31), gave alpha-keto ethers (3a-f) and benzil (4) in good to moderate yields. alpha-Keto ether and benzil formation was very much dependent on the reaction time, the degree of fluorination of the alcohols, and whether a solvent such as CH3CN, DMF or DMA was utilized. In solution, alpha-keto ethers and benzil did not form. Interestingly, under neat conditions, nonfluorinated alcohols, ROH (R = CH3-, CH3CH2-, CH3CH2CH2-, CH3CH2CH2CH2-, CH3CH2CH2CH2CH2CH2-) (5g-k) did not react with trans-stilbene in the presence of MEC-31 but gave 6,6'-dialkoxy-2,2'-bipyridines (6g-k), regioselectively, in excellent isolated yields. On the other hand, fluorinated alcohols did not react with MEC-31. Reaction of MEC-31 with an excess of diethylene glycol (7) gave the bipyridine derivative (8) in 88% isolated yield. Reaction of 8 either with diethylaminosulfur trifluoride (DAST) or bis(2-methoxyethyl)aminosulfur trifluoride (Deoxofluor) readily produced the corresponding difluoro derivative (9) in 85% isolated yield. All new compounds have been characterized by spectroscopic and elemental analysis.
Sigma-Aldrich
2,2,3,3,3-Pentafluoro-1-propanol, 97%