MilliporeSigma
  • TGF-β-induced growth inhibition in B-cell lymphoma correlates with Smad1/5 signalling and constitutively active p38 MAPK.
TGF-β-induced growth inhibition in B-cell lymphoma correlates with Smad1/5 signalling and constitutively active p38 MAPK.
BMC immunology (2010-11-26)
Maren Bakkebø, Kanutte Huse, Vera I Hilden, Erlend B Smeland, Morten P Oksvold
ABSTRACT
Cytokines of the transforming growth factor β (TGF-β) superfamily exert effects on proliferation, apoptosis and differentiation in various cell types. Cancer cells frequently acquire resistance to the anti-proliferative signals of TGF-β, which can be due to mutations in proteins of the signalling cascade. We compared the TGF-β-related signalling properties in B-cell lymphoma cell lines that were sensitive or resistant to TGF-β-induced anti-proliferative effects. TGF-β sensitive cell lines expressed higher cell surface levels of the activin receptor-like kinase 5 (Alk-5), a TGF-β receptor type 1. The expression levels of the other TGF-β and bone morphogenetic protein receptors were comparable in the different cell lines. TGF-β-induced phosphorylation of Smad2 was similar in TGF-β sensitive and resistant cell lines. In contrast, activation of Smad1/5 was restricted to cells that were sensitive to growth inhibition by TGF-β. Moreover, with activin A we detected limited anti-proliferative effects, strong phosphorylation of Smad2, but no Smad1/5 phosphorylation. Up-regulation of the TGF-β target genes Id1 and Pai-1 was identified in the TGF-β sensitive cell lines. Constitutive phosphorylation of MAPK p38 was restricted to the TGF-β sensitive cell lines. Inhibition of p38 MAPK led to reduced sensitivity to TGF-β. We suggest that phosphorylation of Smad1/5 is important for the anti-proliferative effects of TGF-β in B-cell lymphoma. Alk-5 was highly expressed in the sensitive cell lines, and might be important for signalling through Smad1/5. Our results indicate a role for p38 MAPK in the regulation of TGF-β-induced anti-proliferative effects.
Sigma-Aldrich
FR180204, ≥98% (HPLC)