MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mycobacterium indicus pranii supernatant induces apoptotic cell death in mouse peritoneal macrophages in vitro.
Mycobacterium indicus pranii supernatant induces apoptotic cell death in mouse peritoneal macrophages in vitro.
PloS one (2011-02-25)
Rajeev Kumar Pandey, Kunal H Bhatt, Yogesh Dahiya, Ajit Sodhi
ABSTRACT
Mycobacterium indicus pranii (MIP), also known as Mw, is a saprophytic, non-pathogenic strain of Mycobacterium and is commercially available as a heat-killed vaccine for leprosy and recently tuberculosis (TB) as part of MDT. In this study we provide evidence that cell-free supernatant collected from original MIP suspension induces rapid and enhanced apoptosis in mouse peritoneal macrophages in vitro. It is demonstrated that the MIP cell-free supernatant induced apoptosis is mitochondria-mediated and caspase independent and involves mitochondrial translocation of Bax and subsequent release of AIF and cytochrome c from the mitochondria. Experiments with pharmacological inhibitors suggest a possible role of PKC in mitochondria-mediated apoptosis of macrophages.
Sigma-Aldrich
Annexin V-Cy3 Apoptosis Detection Kit