MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 5-Phosphonomethylquinoxalinediones as competitive NMDA receptor antagonists with a preference for the human 1A/2A, rather than 1A/2B receptor composition.
5-Phosphonomethylquinoxalinediones as competitive NMDA receptor antagonists with a preference for the human 1A/2A, rather than 1A/2B receptor composition.
Bioorganic & medicinal chemistry letters (2002-03-23)
Yves P Auberson, Hans Allgeier, Serge Bischoff, Kurt Lingenhoehl, Robert Moretti, Markus Schmutz
ABSTRACT
NMDA antagonists derived from 5-phosphonomethyl-1,4-dihydroquinoxaline-2,3-dione (3a) are potent anticonvulsant agents, and display strong protective effects in the electroshock-induced convulsion assay in mice. Their preference for the human NMDAR 1A/2A over 1A/2B subunit composition was optimized, leading to (1RS,1'S)-PEAQX (9r), which shows a >100-fold selectivity.
Sigma-Aldrich
PEAQX tetrasodium hydrate, ≥98% (HPLC)