MilliporeSigma
  • Nerve growth factor reduces amiloride-sensitive Na+ transport in human airway epithelial cells.
Nerve growth factor reduces amiloride-sensitive Na+ transport in human airway epithelial cells.
Physiological reports (2014-10-28)
Michael J Shimko, Eric J Zaccone, Janet A Thompson, Diane Schwegler-Berry, Michael L Kashon, Jeffrey S Fedan
ABSTRACT
Nerve growth factor (NGF) is overexpressed in patients with inflammatory lung diseases, including virus infections. Airway surface liquid (ASL), which is regulated by epithelial cell ion transport, is essential for normal lung function. No information is available regarding the effect of NGF on ion transport of airway epithelium. To investigate whether NGF can affect ion transport, human primary air-interface cultured epithelial cells were placed in Ussing chambers to obtain transepithelial voltage (-7.1 ± 3.4 mV), short-circuit current (Isc, 5.9 ± 1.0 μA), and transepithelial resistance (750 Ω·cm(2)), and to measure responses to ion transport inhibitors. Amiloride (apical, 3.5 × 10(-5) mol/L) decreased Isc by 55.3%. Apically applied NGF (1 ng/mL) reduced Isc by 5.3% in 5 min; basolaterally applied NGF had no effect. The response to amiloride was reduced (41.6%) in the presence of NGF. K-252a (10 nmol/L, apical) did not itself affect Na(+) transport, but it attenuated the NGF-induced reduction in Na(+) transport, indicating the participation of the trkA receptor in the NGF-induced reduction in Na(+) transport. PD-98059 (30 μmol/L, apical and basolateral) did not itself affect Na(+) transport, but attenuated the NGF-induced reduction in Na(+) transport, indicating that trkA activated the Erk 1/2 signaling cascade. NGF stimulated phosphorylation of Erk 1/2 and the β-subunit of ENaC. K-252a and PD-98059 inhibited these responses. NGF had no effect on Isc in the presence of apical nystatin (50 μmol/L). These results indicate that NGF inhibits Na(+) transport through a trkA-Erk 1/2-activated signaling pathway linked to ENaC phosphorylation.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-β-Tubulin−FITC antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture