MilliporeSigma
  • Sustained activation of sphingomyelin synthase by 2-hydroxyoleic acid induces sphingolipidosis in tumor cells.
Sustained activation of sphingomyelin synthase by 2-hydroxyoleic acid induces sphingolipidosis in tumor cells.
Journal of lipid research (2013-03-09)
Maria Laura Martin, Gerhard Liebisch, Stefan Lehneis, Gerd Schmitz, María Alonso-Sande, Joan Bestard-Escalas, Daniel H Lopez, José Manuel García-Verdugo, Mario Soriano-Navarro, Xavier Busquets, Pablo V Escribá, Gwendolyn Barceló-Coblijn
ABSTRACT
The mechanism of action of 2-hydroxyoleic acid (2OHOA), a potent antitumor drug, involves the rapid and specific activation of sphingomyelin synthase (SMS), leading to a 4-fold increase in SM mass in tumor cells. In the present study, we investigated the source of the ceramides required to sustain this dramatic increase in SM. Through radioactive and fluorescent labeling, we demonstrated that sphingolipid metabolism was altered by a 24 h exposure to 2OHOA, and we observed a consistent increase in the number of lysosomes and the presence of unidentified storage materials in treated cells. Mass spectroscopy revealed that different sphingolipid classes accumulated in human glioma U118 cells after exposure to 2OHOA, demonstrating a specific effect on C16-, C20-, and C22-containing sphingolipids. Based on these findings, we propose that the demand for ceramides required to sustain the SMS activation (ca. 200-fold higher than the basal level) profoundly modifies both sphingolipid and phospholipid metabolism. As the treatment is prolonged, tumor cells fail to adequately metabolize sphingolipids, leading to a situation resembling sphingolipidosis, whereby cell viability is compromised.
Product Description
Avanti
2-OHOA, 2-hydroxyoleic acid (sodium salt), powder
Sigma-Aldrich
2OHOA, ≥98% (HPLC)