MilliporeSigma
Sphingolipids and ceramides in human aqueous humor.
Molecular vision (2013-09-27)
Ayman J Aljohani, Gustavo C Munguba, Yenifer Guerra, Richard K Lee, Sanjoy K Bhattacharya
ABSTRACT
To determine the differential profiles of sphingomyelin, sphingoid base, sphingoid base-1-phosphate and ceramide lipid species and their quantitative differences between control and glaucomatous aqueous humor (AQH) derived from human donors. AQH from control and primary open-angle glaucoma donors was collected and subjected to lipid extraction using suitable modifications of the Bligh and Dyer method. Proteins were estimated using Bradford's method. Lipids were identified and ratiometrically quantified in a two-step process using precursor ion scan or neutral loss scan (NLS) with appropriate class-specific lipid standards on a TSQ Quantum Access Max mass spectrometer following established procedures. Primary human trabecular meshwork cells and video microscopic imaging were used to assess changes in cell shape and motility upon exposure to 20 pmol of Cer(d18:0/18:1(9Z)) in 10% dimethyl sulfoxide (vehicle). We identified several species of sphingomyelin, sphingoid base, sphingoid base-1-phosphate, and ceramides that were common between control and glaucomatous AQH. Some unique lipid species in these classes were also identified in controls but not in glaucoma and vice versa. We found exposure to 20 pmol of Cer(d18:0/18:1(9Z)) resulted in changes in the trabecular meshwork cell shape and observed motility changes compared to vehicle-only control. Most lipids belonging to the sphingomyelin, sphingoid base, sphingoid base-1-phosphate, and ceramide species were common between control and primary open-angle glaucoma donors. However, some sphingolipids and ceramides were found to be uniquely present in control but absent in the glaucomatous AQH and vice versa. Identification of unique lipid species present or absent in the pathophysiological context may contribute further insight into glaucoma pathology.
Product Description
Avanti
18:1-d9 SM, Avanti Research - A Croda Brand