MilliporeSigma
  • Autoantibodies and gastrointestinal symptoms in infertile women in relation to in vitro fertilization.
Autoantibodies and gastrointestinal symptoms in infertile women in relation to in vitro fertilization.
BMC pregnancy and childbirth (2013-11-07)
Oskar Hammar, Bodil Roth, Mariette Bengtsson, Thomas Mandl, Bodil Ohlsson
ABSTRACT
Prior reports suggest a link between gonadotropin-releasing hormone (GnRH) and gastrointestinal function. The aim of the study was to prospectively investigate women subjected to in vitro fertilization (IVF) using the GnRH analog buserelin, taking into account gastrointestinal symptoms and antibody development against buserelin, GnRH, luteinizing hormone (LH), and their receptors. Gastrointestinal symptoms were registered by the Visual Analogue Scale for Irritable Bowel Syndrome (VAS-IBS) before and after IVF treatment, and five years later. Health-related quality of life was evaluated by the 36-item Short-Form questionnaire (SF-36). ELISA was used for antibody analyses before and after treatment. Data were compared with women from the general population. In total, 124 patients were investigated before and after IVF, and 62 were re-evaluated after five years. Buserelin treatment led to significant impairment of constipation (p = 0.004), nausea and vomiting (p = 0.035), psychological well-being (p = 0.000), and the intestinal symptoms' influence on daily life (p = 0.027). At 5-year follow-up, abdominal pain was worsened (p = 0.041), but psychological well-being was improved (p = 0.036), compared to prior treatment, and 15% had an observable deterioration in gastrointestinal symptoms. None developed severe dysmotility. Patients had higher prevalence of IgG antibodies against LH (p = 0.001) and its receptor (p = 0.016), and IgM antibodies against the GnRH receptor (p = 0.001) prior treatment compared with controls, but no antibody development was observed after IVF. Patients experience gastrointestinal symptoms during buserelin treatment, and abdominal pain is still increased after five years, but buserelin does not increase antibody formation against GnRH, LH or their receptors.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution