MilliporeSigma
  • Age- and light-dependent development of localised retinal atrophy in CCL2(-/-)CX3CR1(GFP/GFP) mice.
Age- and light-dependent development of localised retinal atrophy in CCL2(-/-)CX3CR1(GFP/GFP) mice.
PloS one (2013-05-03)
Mei Chen, Jose R Hombrebueno, Chang Luo, Rosana Penalva, Jiawu Zhao, Liza Colhoun, Sudha Pirya Soundara Pandi, John V Forrester, Heping Xu
ABSTRACT
Previous studies have shown that CCL2/CX3CR1 deficient mice on C57BL/6N background (with rd8 mutation) have an early onset (6 weeks) of spontaneous retinal degeneration. In this study, we generated CCL2(-/-)CX3CR1(GFP/GFP) mice on the C57BL/6J background. Retinal degeneration was not detected in CCL2(-/-)CX3CR1(GFP/GFP) mice younger than 6 months. Patches of whitish/yellowish fundus lesions were observed in 17∼60% of 12-month, and 30∼100% of 18-month CCL2(-/-)CX3CR1(GFP/GFP) mice. Fluorescein angiography revealed no choroidal neovascularisation in these mice. Patches of retinal pigment epithelium (RPE) and photoreceptor damage were detected in 30% and 50% of 12- and 18-month CCL2(-/-)CX3CR1(GFP/GFP) mice respectively, but not in wild-type mice. All CCL2(-/-)CX3CR1(GFP/GFP) mice exposed to extra-light (∼800lux, 6 h/day, 6 months) developed patches of retinal atrophy, and only 20-25% of WT mice which underwent the same light treatment developed atrophic lesions. In addition, synaptophysin expression was detected in the outer nucler layer (ONL) of area related to photoreceptor loss in CCL2(-/-)CX3CR1(GFP/GFP) mice. Markedly increased rhodopsin but reduced cone arrestin expression was observed in retinal outer layers in aged CCL2(-/-)CX3CR1(GFP/GFP) mice. GABA expression was reduced in the inner retina of aged CCL2(-/-)CX3CR1(GFP/GFP) mice. Significantly increased Müller glial and microglial activation was observed in CCL2(-/-)CX3CR1(GFP/GFP) mice compared to age-matched WT mice. Macrophages from CCL2(-/-)CX3CR1(GFP/GFP) mice were less phagocytic, but expressed higher levels of iNOS, IL-1β, IL-12 and TNF-α under hypoxia conditions. Our results suggest that the deletions of CCL2 and CX3CR1 predispose mice to age- and light-mediated retinal damage. The CCL2/CX3CR1 deficient mouse may thus serve as a model for age-related atrophic degeneration of the RPE, including the dry type of macular degeneration, geographic atrophy.
Sigma-Aldrich
Anti-GABA Antibody, serum, Chemicon®