MilliporeSigma
  • Two functionally distinct isoforms of TL1A (TNFSF15) generated by differential ectodomain shedding.
Two functionally distinct isoforms of TL1A (TNFSF15) generated by differential ectodomain shedding.
The journals of gerontology. Series A, Biological sciences and medical sciences (2010-08-03)
Christoph Mück, Dietmar Herndler-Brandstetter, Lucia Micutkova, Beatrix Grubeck-Loebenstein, Pidder Jansen-Dürr
ABSTRACT
Tumor necrosis factor-like cytokine 1A (TL1A) is expressed in endothelial cells and contributes to T-cell activation, via an extracellular fragment TL1A(L72-L251), generated by ectodomain shedding. Fragments of TL1A, referred to as vascular endothelial growth inhibitor, were found to induce growth arrest and apoptosis in endothelial cells; however, the underlying mechanisms remained obscure. Here, we show that full-length TL1A is the major detectable gene product in both human umbilical vein endothelial cells and circulating endothelial progenitor cells. TL1A expression was significantly enhanced in senescent circulating endothelial progenitor cells, and knockdown of TL1A partially reverted senescence. TL1A overexpression induced premature senescence in both circulating endothelial progenitor cells and human umbilical vein endothelial cells. We also identified a novel extracellular fragment of TL1A, TL1A(V84-L251), resulting from differential ectodomain shedding, which induced growth arrest and apoptosis in human umbilical vein endothelial cells. These findings suggest that TL1A is involved in the regulation of endothelial cell senescence, via a novel fragment produced by differential ectodomain shedding.
Sigma-Aldrich
TAPI-1, TAPI-1, CAS 171235-71-5, is a structural analog of TAPI-0 with similar in vitro efficacy for the inhibition of MMPs and TACE. Blocks the shedding of several cell surface proteins.
Sigma-Aldrich
Glycoprotein Deglycosylation Kit