MilliporeSigma
  • Cloning and characterization of a glucosyltransferase from Crocus sativus stigmas involved in flavonoid glucosylation.
Cloning and characterization of a glucosyltransferase from Crocus sativus stigmas involved in flavonoid glucosylation.
BMC plant biology (2009-08-22)
Angela Rubio Moraga, Almudena Trapero Mozos, Oussama Ahrazem, Lourdes Gómez-Gómez
ABSTRACT
Flavonol glucosides constitute the second group of secondary metabolites that accumulate in Crocus sativus stigmas. To date there are no reports of functionally characterized flavonoid glucosyltransferases in C. sativus, despite the importance of these compounds as antioxidant agents. Moreover, their bitter taste makes them excellent candidates for consideration as potential organoleptic agents of saffron spice, the dry stigmas of C. sativus. Using degenerate primers designed to match the plant secondary product glucosyltransferase (PSPG) box we cloned a full length cDNA encoding CsGT45 from C. sativus stigmas. This protein showed homology with flavonoid glucosyltransferases. In vitro reactions showed that CsGT45 catalyses the transfer of glucose from UDP_glucose to kaempferol and quercetin. Kaempferol is the unique flavonol present in C. sativus stigmas and the levels of its glucosides changed during stigma development, and these changes, are correlated with the expression levels of CsGT45 during these developmental stages. Findings presented here suggest that CsGT45 is an active enzyme that plays a role in the formation of flavonoid glucosides in C. sativus.
Sigma-Aldrich
Kaempferol 7-O-β -D-glucopyranoside, ≥90.0% (HPLC)