MilliporeSigma
  • MicroRNA regulation of Cbx7 mediates a switch of Polycomb orthologs during ESC differentiation.
MicroRNA regulation of Cbx7 mediates a switch of Polycomb orthologs during ESC differentiation.
Cell stem cell (2012-01-10)
Ana O'Loghlen, Ana M Muñoz-Cabello, Alexandre Gaspar-Maia, Hsan-Au Wu, Ana Banito, Natalia Kunowska, Tomas Racek, Helen N Pemberton, Patrizia Beolchi, Fabrice Lavial, Osamu Masui, Michiel Vermeulen, Thomas Carroll, Johannes Graumann, Edith Heard, Niall Dillon, Veronique Azuara, Ambrosius P Snijders, Gordon Peters, Emily Bernstein, Jesus Gil
ABSTRACT
The Polycomb Group (PcG) of chromatin modifiers regulates pluripotency and differentiation. Mammalian genomes encode multiple homologs of the Polycomb repressive complex 1 (PRC1) components, including five orthologs of the Drosophila Polycomb protein (Cbx2, Cbx4, Cbx6, Cbx7, and Cbx8). We have identified Cbx7 as the primary Polycomb ortholog of PRC1 complexes in embryonic stem cells (ESCs). The expression of Cbx7 is downregulated during ESC differentiation, preceding the upregulation of Cbx2, Cbx4, and Cbx8, which are directly repressed by Cbx7. Ectopic expression of Cbx7 inhibits differentiation and X chromosome inactivation and enhances ESC self-renewal. Conversely, Cbx7 knockdown induces differentiation and derepresses lineage-specific markers. In a functional screen, we identified the miR-125 and miR-181 families as regulators of Cbx7 that are induced during ESC differentiation. Ectopic expression of these miRNAs accelerates ESC differentiation via regulation of Cbx7. These observations establish a critical role for Cbx7 and its regulatory miRNAs in determining pluripotency.
Sigma-Aldrich
MISSION® esiRNA, targeting human PPIB