MilliporeSigma
  • Intestinal absorption of forsythoside A in in situ single-pass intestinal perfusion and in vitro Caco-2 cell models.
Intestinal absorption of forsythoside A in in situ single-pass intestinal perfusion and in vitro Caco-2 cell models.
Acta pharmacologica Sinica (2012-07-10)
Wei Zhou, Liu-qing Di, Juan Wang, Jin-jun Shan, Shi-jia Liu, Wen-zheng Ju, Bao-chang Cai
ABSTRACT
To investigate the mechanisms underlying the intestinal absorption of the major bioactive component forsythoside A (FTA) extracted from Forsythiae fructus. An in vitro Caco-2 cell model and a single-pass intestinal perfusion in situ model in SD rats were used. In the in vitro Caco-2 cell model, the mean apparent permeability value (P(app)-value) was 4.15×10(-7) cm/s in the apical-to-basolateral (AP-BL) direction. At the concentrations of 2.6-10.4 μg/mL, the efflux ratio of FTA in the bi-directional transport experiments was approximately 1.00. After the transport, >96% of the apically loaded FTA was retained on the apical side, while >97% of the basolaterally loaded FTA was retained on the basolateral side. The P(app)-values of FTA were inversely correlated with the transepithelial electrical resistance. The paracellular permeability enhancers sodium caprate and EDTA, the P-gp inhibitor verapamil and the multidrug resistance related protein (MRP) inhibitors cyclosporine and MK571 could concentration-dependently increase the Papp-values, while the uptake (OATP) transporter inhibitors diclofenac sodium and indomethacin could concentration-dependently decrease the P(app)-values. The intake transporter SGLT1 inhibitor mannitol did not cause significant change in the P(app)-values. In the in situ intestinal perfusion model, both the absorption rate constant (K(a)) and the effective permeability (P(eff)-values) following perfusion of FTA 2.6, 5.2 and 10.4 μg/mL via the duodenum, jejunum and ileum had no significant difference, although the values were slightly higher for the duodenum as compared to those in the jejunum and ileum. The low, medium and high concentrations of verapamil caused the largest increase in the P(eff)-values for duodenum, jejunum and ileum, respectively. Sodium caprate, EDTA and cyclosporine resulted in concentration-dependent increase in the P(eff)-values. Diclofenac sodium and indomethacin caused concentration-dependent decrease in the Peff-values. Mannitol did not cause significant change in the P(app)-values for the duodenum, jejunum or ileum. The results suggest that the intestinal absorption of FTA may occur through passive diffusion, and the predominant absorption site may be in the upper part of small intestine. Paracellular transport route is also involved. P-gp, MRPs and OATP may participate in the absorption of FTA in the intestine. The low permeability of FTA contributes to its low oral bioavailability.
Sigma-Aldrich
Sodium decanoate, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
MK-571 sodium salt hydrate, ≥95% (HPLC)
Pricing and availability is not currently available.